Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isopropylmagnesium Chloride Lithium Chloride Complex Solution

Catalog No. 6569842L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
2 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-698-42L 2 L
65-698-425ML 4 x 25 mL
65-698-4100ML 100 mL
65-698-4800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 65-698-42L Supplier Sigma Aldrich Supplier No. 6569842L
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used with CH3MgCl/LiCl to affect magnesium-halogen exchange on unprotected aryl carboxylic acids.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

Density 0.951 g/mL at 25°C
Molecular Formula C3H7Cl2LiMg
Linear Formula (CH3)2CHMgCl -+ LiCl
Quantity 2 L
MDL Number MFCD07784514
Synonym Turbo Grignard
Molecular Weight (g/mol) 145.24 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.