Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58536AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58536AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58536AD Supplier Thermo Scientific Chemicals Supplier No. H58536AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 77047-87-1 | C3H7BrZn | 188.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylzinc bromide is employed in croos-coupling reactions.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents,
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77047-87-1
Molecular Formula C3H7BrZn
Molecular Weight (g/mol) 188.37
MDL Number MFCD06200639
InChI Key NTUATXZETUXBSR-UHFFFAOYSA-M
Synonym 2-propylzinc bromide solution 0.5m in t, bromo isopropyl zinc, 2-propylzinc bromide, 0.50 m in thf, 2-propylzinc bromide 0.5 m in tetrahydrofuran, 2-propylzinc bromide solution, 0.5 m in thf, isopropylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 12910424
IUPAC Name bromozincylium; propan-2-ide
SMILES [Zn+]Br.C[CH-]C

Specifications

Density 0.963
Linear Formula (CH3)2CHZnBr
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Reacts with water.
Formula Weight 188.37
Chemical Name or Material Isopropylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.