Learn More
Selleck Chemical LLC Isoproterenol sulfate
SDP
Catalog No. 5000003266
Encompass_Preferred
Isoproterenol Sulfate is the sulfate salt form of isoproterenol a beta-adrenergic receptor agonist with bronchodilator activity Isoproterenol sulfate dihydrate can be used to induce animal models of Angina Pectoris
Catalog No. 50-000-03266 Supplier Selleck Chemical LLC Supplier No. S56831G
Isoproterenol Sulfate is the sulfate salt form of isoproterenol a beta-adrenergic receptor agonist with bronchodilator activity Isoproterenol sulfate dihydrate can be used to induce animal models of Angina Pectoris