Learn More
Apexbio Technology LLC Isoproterenol sulfate dihydrate 299-95-6 25mg
SDP
Catalog No. 5000092695
Encompass_Preferred
Isoproterenol sulfate dihydrate (CAS 299-95-6) is a non-selective -adrenergic receptor agonist that exerts its biological effects by activating both 1- and 2-adrenergic receptors Through this mechanism it induces bronchodilation and peripheral vasodilation In biomedical research isoproterenol sulfate dihydrate is commonly utilized to investigate physiological and pathological processes related to bradycardia bronchial asthma and cardiovascular function enabling the study of -adrenergic signaling and associated responses in various experimental models
Catalog No. 50-000-92695 Supplier Apexbio Technology LLC Supplier No. C640225
Isoproterenol sulfate dihydrate (CAS 299-95-6) is a non-selective -adrenergic receptor agonist that exerts its biological effects by activating both 1- and 2-adrenergic receptors Through this mechanism it induces bronchodilation and peripheral vasodilation In biomedical research isoproterenol sulfate dihydrate is commonly utilized to investigate physiological and pathological processes related to bradycardia bronchial asthma and cardiovascular function enabling the study of -adrenergic signaling and associated responses in various experimental models