Learn More
AdipoGen Isoprothiolane
SDP
Supplier:  AdipoGen CDXI0048GG25
Encompass_Preferred
Chemical. CAS 50512-35-1. Formula C12H18O4S2. MW 290.4. Synthetic. Isoprothiolane is a dithiolane pesticide. Isoprothiolane is commonly used in agriculture as a fungicide and insecticide to control planthoppers and blast disease in rice plants. Shown to regulate mammary epithelial cell growth in vitro by modulation production of cytokines and improving hepatic lipid metabolism in cows. Compound can be used as analytical reference material.
Catalog No. 50-256-0641