Learn More
Cayman Chemical Isoproterenol (hydrochloride), Purity: ≥98%, CAS: 51-30-9, 100 mg
SDP
Catalog No. 502969448
Encompass_Preferred
A β1- and β2-AR agonist (Kis = 224 and 458 nM, respectively); selective for β1- and β2-ARs over β3-ARs (Ki = 1,570 nM); inhibits contractions in isolated field-stimulated rat vas deferens (EC50 = 45.6 nM); decreases blood pressure and increases water intake in nephrectomized rats at 0.33 mg/kg; reduces blood pressure and increases heart rate in renal hypertensive rabbits; inhibits histamine-induced bronchospasms in anesthetized dogs
Catalog No. 50-296-9448 Supplier Cayman Chemical Supplier No. 15592100 MG
A β1- and β2-AR agonist (Kis = 224 and 458 nM, respectively); selective for β1- and β2-ARs over β3-ARs (Ki = 1,570 nM); inhibits contractions in isolated field-stimulated rat vas deferens (EC50 = 45.6 nM); decreases blood pressure and increases water intake in nephrectomized rats at 0.33 mg/kg; reduces blood pressure and increases heart rate in renal hypertensive rabbits; inhibits histamine-induced bronchospasms in anesthetized dogs