Learn More
Medchemexpress LLC Isoproturon-d3 | 352438-80-3 | 99.8% | 1 MG
SDP
Catalog No. 5000412141
Encompass_Preferred
Isoproturon-d3 is the deuterium labeled Isoproturon. Isoproturon belongs to the phenylurea herbicide family and is a systemic and selective herbicide. It is widely applied for killing weeds in farmland, specifically annual grasses and broad-leaved weeds in spring and winter wheat, winter rye, and spring and winter barley. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12141 Supplier Medchemexpress LLC Supplier No. HYB1859S11MG
Isoproturon-d3 is the deuterium labeled Isoproturon. Isoproturon belongs to the phenylurea herbicide family and is a systemic and selective herbicide. It is widely applied for killing weeds in farmland, specifically annual grasses and broad-leaved weeds in spring and winter wheat, winter rye, and spring and winter barley. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.