Learn More
Medchemexpress LLC Isoproturon-d3 | 352438-80-3 | 99.8% | 209.30 | C12H15D3N2O | 10 MG
SDP
Catalog No. 5000298906
Encompass_Preferred
Isoproturon-d3 is the deuterium-labeled analogue of the phenylurea herbicide isoproturon, supplied as a stable isotope reference standard for analytical and research use. It is designed for use as an internal standard or tracer in quantitative analytical methods, particularly LC-MS, to support environmental fate, residue analysis, and metabolic studies. Supplied in a 10 mg vial with documentation for batch verification.
  • Stable deuterium label for accurate quantification in mass spectrometry
  • Suitable as an internal standard for environmental residue and metabolism studies
  • High reported purity for trace-level analysis
  • Provided with supporting documentation such as COA and chromatographic data
  • Compact research-grade quantity compatible with routine analytical workflows
Catalog No. 50-002-98906 Supplier Medchemexpress LLC Supplier No. HYB1859S110MG
Isoproturon-d3 is the deuterium-labeled analogue of the phenylurea herbicide isoproturon, supplied as a stable isotope reference standard for analytical and research use. It is designed for use as an internal standard or tracer in quantitative analytical methods, particularly LC-MS, to support environmental fate, residue analysis, and metabolic studies. Supplied in a 10 mg vial with documentation for batch verification.
  • Stable deuterium label for accurate quantification in mass spectrometry
  • Suitable as an internal standard for environmental residue and metabolism studies
  • High reported purity for trace-level analysis
  • Provided with supporting documentation such as COA and chromatographic data
  • Compact research-grade quantity compatible with routine analytical workflows