Learn More
Medchemexpress LLC Isoproturon-d3 | 352438-80-3 | 99.8% | 5 MG
SDP
Catalog No. 5000412142
Encompass_Preferred
Isoproturon-d3 is a deuterium-labeled form of Isoproturon, a systemic and selective phenylurea herbicide. It is widely used to control annual grasses and broad-leaved weeds in spring and winter wheat, winter rye, and spring and winter barley. This compound can be utilized as a tracer and an internal standard for quantitative analysis.
  • Deuterium labeled compound.
  • Serves as a tracer for quantitation during drug development.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Functions as a systemic and selective herbicide.
  • Aids in the control of annual grasses and broad-leaved weeds in various crops.
Catalog No. 50-004-12142 Supplier Medchemexpress LLC Supplier No. HYB1859S15MG
Isoproturon-d3 is a deuterium-labeled form of Isoproturon, a systemic and selective phenylurea herbicide. It is widely used to control annual grasses and broad-leaved weeds in spring and winter wheat, winter rye, and spring and winter barley. This compound can be utilized as a tracer and an internal standard for quantitative analysis.
  • Deuterium labeled compound.
  • Serves as a tracer for quantitation during drug development.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Functions as a systemic and selective herbicide.
  • Aids in the control of annual grasses and broad-leaved weeds in various crops.