Learn More
Cayman Chemical IsopteropodIn 10mg
SDP
Catalog No. 502978400
Encompass_Preferred
An oxindole alkaloid with diverse biological activities; induces activation of PXR in a reporter assay using DPX2 cells expressing the human receptor (EC50 = 0.7656 µM); a positive modulator of M1 mAChRs and 5-HT2 receptor (EC50s = 9.92 and 14.5 µM for the rat receptors, respectively); active against S. aureus and B. subtilis (MICs = 408 and 679 µM, respectively); inhibits the proliferation of CCRF CEM C7H2 lymphoblastic leukemia cells at 100 µM
Catalog No. 50-297-8400 Supplier Cayman Chemical Supplier No. 4023710 MG
An oxindole alkaloid with diverse biological activities; induces activation of PXR in a reporter assay using DPX2 cells expressing the human receptor (EC50 = 0.7656 µM); a positive modulator of M1 mAChRs and 5-HT2 receptor (EC50s = 9.92 and 14.5 µM for the rat receptors, respectively); active against S. aureus and B. subtilis (MICs = 408 and 679 µM, respectively); inhibits the proliferation of CCRF CEM C7H2 lymphoblastic leukemia cells at 100 µM