Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Isopulegol

Catalog No. 4390531ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-905-31ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 43-905-31ML Supplier Sigma Aldrich Supplier No. 4390531ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isopulegol is a monoterpene alcohol, which is generally found in essential oils of various plants. It is widely used as a fragrance ingredient in cosmetics, shampoos and toilet soaps. Isopulegol is also a key intermediate in the synthesis of (-)-menthol.(+)-Isopulegol can be used as a starting material to synthesize:A natural product (-)-isopiperitenone.Medicinally important octahydro-2H-chromen-4-ol derivatives by reacting with benzaldehydes.Antituberculosis diterpenoid named 12-epi-ileabethoxazole.

Specifications

CAS 10365-98-7
Melting Point 160°C to 163°C
Molecular Formula C7H9BO3
Refractive Index n20/D 1.469
Linear Formula CH3OC6H4B(OH)2
MDL Number MFCD00161359
Quantity 1 mL
Synonym (m-Methoxyphenyl)boronic acid; 3-Methoxybenzeneboronic acid; m-Anisylboronic acid; m-Methoxybenzeneboronic acid
Molecular Weight (g/mol) 151.96 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.