Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Isopulegol

Catalog No. 4390611ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-906-11ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 43-906-11ML Supplier Sigma Aldrich Supplier No. 4390611ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isopulegol is monoterpene alcohol, a useful ingredient for the production of fragrances in perfume industries. It is also used as a starting material in the manufacture of menthol by the hydrogenation process. Menthol is an important component in cosmetics, pharmaceuticals, and toothpaste.(-)-Isopulegol can be used as a starting material for the enantioselective preparation of: 8-arylmenthols by Smiles-Truce rearrangement of aryl sulfonates.,-+ Stereoisomers of 5,9-dimethylpentadecane. Octahydro-2H-chromen-4-ol by Prins cyclization with vanillin in the presence of montmorillonite clay as the catalyst p-menthane-3,8,9-triol by catalytic Sharpless dihydroxylation.

Specifications

CAS 89598-96-9
Melting Point 164°C to 168°C
Molecular Formula C6H6BBrO2
Refractive Index n20/D 1.471
Linear Formula BrC6H4B(OH)2
MDL Number MFCD00239386
Quantity 5 g
Synonym 3-Bromobenzeneboronic acid
Molecular Weight (g/mol) 200.83 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.