Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Isopulegol | 89-79-2 | 99.8% | 100 G
SDP

Catalog No. 5000233471
Encompass_Preferred

Isopulegol is a chiral monoterpene alcohol used as a research reagent and intermediate in antiviral and synthetic chemistry studies. It is supplied in laboratory pack sizes and used as a building block for further chemical transformations.

  • Chiral monoterpene alcohol suitable for research applications.
  • Cas number 89-79-2 for unambiguous identification.
  • Purity typically around 99.8% (reported values vary by batch).
  • Molecular weight 154.25 g/mol.
  • Soluble in DMSO for experimental use.
  • Available in 100 G pack sizes for synthesis and scale-up.

Catalog No. 50-002-33471 Supplier Medchemexpress LLC Supplier No. HY121206100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopulegol is a chiral monoterpene alcohol used as a research reagent and intermediate in antiviral and synthetic chemistry studies. It is supplied in laboratory pack sizes and used as a building block for further chemical transformations.

  • Chiral monoterpene alcohol suitable for research applications.
  • Cas number 89-79-2 for unambiguous identification.
  • Purity typically around 99.8% (reported values vary by batch).
  • Molecular weight 154.25 g/mol.
  • Soluble in DMSO for experimental use.
  • Available in 100 G pack sizes for synthesis and scale-up.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.