Learn More
Medchemexpress LLC (-)-Isopulegol | 89-79-2 | 99.8% | 25 G
SDP
Catalog No. 5000231769
Encompass_Preferred
(-)-Isopulegol is a monoterpene alcohol used in biochemical and antiviral research. It is supplied as a high-purity research reagent for laboratory use only and is commonly available in small pack sizes, including 25 g.
  • Monoterpene alcohol with CAS 89-79-2.
  • Chemical formula C10H18O and molecular weight 154.25.
  • High purity as confirmed by COA (99.83% by GC).
  • Colorless, clear liquid with a mint-like odor.
  • Intended for research use only, not for human or veterinary use.
  • Suitable for biochemical and antiviral activity studies.
Catalog No. 50-002-31769 Supplier Medchemexpress LLC Supplier No. HY12120625G
(-)-Isopulegol is a monoterpene alcohol used in biochemical and antiviral research. It is supplied as a high-purity research reagent for laboratory use only and is commonly available in small pack sizes, including 25 g.
  • Monoterpene alcohol with CAS 89-79-2.
  • Chemical formula C10H18O and molecular weight 154.25.
  • High purity as confirmed by COA (99.83% by GC).
  • Colorless, clear liquid with a mint-like odor.
  • Intended for research use only, not for human or veterinary use.
  • Suitable for biochemical and antiviral activity studies.