Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopulegol (mixture of isomers) 90.0+%, TCI America™
SDP

Catalog No. M032025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M032025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M032025ML Supplier TCI America Supplier No. M032025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7786-67-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00134655
InChI Key ZYTMANIQRDEHIO-UHFFFAOYNA-N
Synonym p-menth-8-en-3-ol, isopulegol, 8 9-p-menthen-3-ol, 5-methyl-2-prop-1-en-2-yl cyclohexanol, cyclohexanol, 5-methyl-2-1-methylethenyl, 1-methyl-4-isopropenylcyclohexan-3-ol, dl-isopulegol, p-8 9-menthen-3-ol, 5-methyl-2-1-methylvinyl cyclohexan-1-ol, 5-methyl-2-prop-1-en-2-yl cyclohexan-1-ol
PubChem CID 24585
IUPAC Name 5-methyl-2-(prop-1-en-2-yl)cyclohexan-1-ol
SMILES CC1CCC(C(O)C1)C(C)=C

Specifications

Quantity 25 mL
Formula Weight 154.25
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopulegol (mixture of isomers)
EINECSNumber (3)-2334
RTECSNumber OT0190000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.