missing translation for 'onlineSavingsMsg'
Learn More

Isopulegol, MP Biomedicals

Catalog No. ICN214076 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214076 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214076 Supplier MP Biomedicals, Inc Supplier No. 0521407601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89-79-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00134655
InChI Key ZYTMANIQRDEHIO-UHFFFAOYNA-N
Synonym isopulegol, --isopulegol, l-isopulegol, --l-isopulegol, coolact p, unii-3th92o3bxn, cyclohexanol, 5-methyl-2-1-methylethenyl-, 1r,2s,5r, 3th92o3bxn, iso-pulegol, isopulegol natural
PubChem CID 170833
ChEBI CHEBI:70479
IUPAC Name 5-methyl-2-(prop-1-en-2-yl)cyclohexan-1-ol
SMILES CC1CCC(C(O)C1)C(C)=C

Specifications

Quantity 100 g
Formula Weight 154.25
Chemical Name or Material Isopulegol
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.