Learn More
TARGETMOL CHEMICALS INC ISOQUERCETIN 25MG
SDP
Catalog No. 502579718
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Isoquercetin (3-Glucosylquercetin) is a potential stimulator of bone mineralization used for prophylaxis of osteoporotic disorders. 2. Isoquercetin may be as a potential therapeutic agent against neurodegeneration in Parkinson's disease. 3. Isoquercetin is an inhibitor of Wnt/(beta)-catenin and may be as a potential novel anti-tumoral agent such as against human pancreatiliver cancer related to opioid receptors and to the activation of the mitogen-activated protein kinase (MAPK) signalling pathway. . purity: 99%
Catalog No. 50-257-9718 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S075425MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Isoquercetin (3-Glucosylquercetin) is a potential stimulator of bone mineralization used for prophylaxis of osteoporotic disorders. 2. Isoquercetin may be as a potential therapeutic agent against neurodegeneration in Parkinson's disease. 3. Isoquercetin is an inhibitor of Wnt/(beta)-catenin and may be as a potential novel anti-tumoral agent such as against human pancreatiliver cancer related to opioid receptors and to the activation of the mitogen-activated protein kinase (MAPK) signalling pathway. . purity: 99%