Learn More
Cayman Chemical IsoquercItrosIde 10mg
SDP
Supplier:  Cayman Chemical 2080210 MG
Encompass_Preferred
A 3-O-glucoside of quercetin that displays antioxidant and chemoprotective effects both in vitro and in vivo.
Catalog No. 50-298-3675