Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinolin-3-amine, 97%, Thermo Scientific™

Catalog No. BTB10019EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB10019EA 10 g Amber Glass Bottle
BTB10019CB 250 mg Amber Glass Bottle
BTB10019DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB10019EA Supplier Thermo Scientific Chemicals Supplier No. BTB10019EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25475-67-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
InChI Key VYCKDIRCVDCQAE-UHFFFAOYSA-N
Synonym 3-aminoisoquinoline, 3-isoquinolinamine, isoquinolin-3-ylamine, isoquinoline, 3-amino, 3-isoquinolylamine, isoquinoline-3-amine, chembl61743, 2h-isoquinolin-3-imine, isoquinolin-3-imine, zlchem 309
PubChem CID 311869
IUPAC Name isoquinolin-3-amine
SMILES C1=CC=C2C=NC(=CC2=C1)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 144.18
Percent Purity 97%
Chemical Name or Material Isoquinolin-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.