Learn More
Medchemexpress LLC Isoquinoline, 3,4-dihydro-1-phenyl- | 52250-50-7 | ≥96.0% | 207.28 | 5 G
SDP
Catalog No. 5000401217
Encompass_Preferred
1-Phenyl-3,4-dihydroisoquinoline is a chemical compound with the molecular formula C15H13N and a molecular weight of 207.28. This product appears as a liquid, ranging from light yellow to brown in color, and is confirmed to have a purity of at least 96.0% based on NMR analysis.
  • Identified by CAS number 52250-50-7
  • Minimum purity of 96.0%
  • Consistent chemical structure confirmed by ¹H NMR spectrum
  • Stable for extended periods when stored properly, with specific conditions for pure form and in solvent
Catalog No. 50-004-01217 Supplier Medchemexpress LLC Supplier No. HY321365G
1-Phenyl-3,4-dihydroisoquinoline is a chemical compound with the molecular formula C15H13N and a molecular weight of 207.28. This product appears as a liquid, ranging from light yellow to brown in color, and is confirmed to have a purity of at least 96.0% based on NMR analysis.
  • Identified by CAS number 52250-50-7
  • Minimum purity of 96.0%
  • Consistent chemical structure confirmed by ¹H NMR spectrum
  • Stable for extended periods when stored properly, with specific conditions for pure form and in solvent