Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinoline-3-carboxylic Acid Monohydrate 98.0+%, TCI America™
SDP

Catalog No. I05095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05095G 5 g
I05091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05095G Supplier TCI America Supplier No. I05095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 203626-75-9 | C10H9NO3 | 191.186 g/mol

Chemical Identifiers

CAS 203626-75-9
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00075137
InChI Key IXFLTVWYYQIVRB-UHFFFAOYSA-N
Synonym isoquinoline-3-carboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate, isoquinoline-3-carboxylic acid monohydrate, isoquinoline-3-carboxylic acid hydrate 1:x, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, pubchem10167, acmc-1clhb, 3-isoquinolinecarboxylicacid hydrate, isoquinoline 3-carboxylic acid hydrate
PubChem CID 2736704
IUPAC Name isoquinoline-3-carboxylic acid;hydrate
SMILES C1=CC=C2C=NC(=CC2=C1)C(=O)O.O

Specifications

Melting Point 167°C
Quantity 5 g
Formula Weight 173.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline-3-carboxylic Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.