Learn More
Medchemexpress LLC Isoquinoline, 5-(1-piperazinyl)-, hydrochloride (1:1) | 209733-17-5 | 99.6% | 249.74 | 250 MG
SDP
Catalog No. 5000402910
Encompass_Preferred
This product is described as a yellow to orange solid. It has been tested and complies with the given specifications.
  • Suitable for storage at 4°C, sealed, and away from moisture.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture.
Catalog No. 50-004-02910 Supplier Medchemexpress LLC Supplier No. 250MGHY23703A
This product is described as a yellow to orange solid. It has been tested and complies with the given specifications.
  • Suitable for storage at 4°C, sealed, and away from moisture.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture.