Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinoline-5-carboxaldehyde, 97%

Catalog No. AAL19636MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19636MD 250 mg
AAL1963603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19636MD Supplier Thermo Scientific Chemicals Supplier No. L19636MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 80278-67-7 | C10H7NO | 157.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80278-67-7
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD03412483
InChI Key ILRSABOCKMOFGW-UHFFFAOYSA-N
Synonym 5-isoquinolinecarboxaldehyde, isoquinoline-5-carboxaldehyde, 5-formylisoquinoline, ksc447q7n, 5-isoquinolinecarbaldehyde, 5-isoquinolinecarboxaldehyde 6ci,9ci
PubChem CID 7016853
IUPAC Name isoquinoline-5-carbaldehyde
SMILES C1=CC2=C(C=CN=C2)C(=C1)C=O

Specifications

Melting Point 113°C to 116°C
Quantity 250 mg
Beilstein 112875
Sensitivity Air Sensitive
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material Isoquinoline-5-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.