Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinoline-5-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. I09425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09425G 5 g
I09421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09425G Supplier TCI America Supplier No. I09425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80278-67-7
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD03412483
InChI Key ILRSABOCKMOFGW-UHFFFAOYSA-N
Synonym 5-isoquinolinecarboxaldehyde, isoquinoline-5-carboxaldehyde, 5-formylisoquinoline, ksc447q7n, 5-isoquinolinecarbaldehyde, 5-isoquinolinecarboxaldehyde 6ci,9ci
PubChem CID 7016853
IUPAC Name isoquinoline-5-carbaldehyde
SMILES C1=CC2=C(C=CN=C2)C(=C1)C=O

Specifications

Melting Point 119°C
Color Yellow
Quantity 5 g
Formula Weight 157.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline-5-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.