Learn More
Medchemexpress LLC HY-W012732 500mg Medchemexpress, Isoquinoline CAS:119-65-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW012732500MG
Encompass_Preferred
Medchemexpress, HY-W012732 500mg Isoquinoline CAS:119-65-3 Isoquinoline is an analog of pyridine. Isoquinoline structural-based alkaloids, such as tropoloisoquinoline, phthalideisoquinoline, and naphthylisoquinoline has anti-cancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3337