Learn More
Description
- 1-phenyl-2-butanone was used in the preparation of 1-bromo-1-phenyl-2-butanone
- It was used as solute to evaluate the hydroxyl group-solvent and carbonyl group-solvent specific interactions in acetonitrile/water mixtures using the alltima c18 stationary phase by HPLC
Specifications
Specifications
| Molecular Formula | C9H8BNO2 |
| Linear Formula | C9H8BNO2 |
| MDL Number | MFCD10697418 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 172.98 |
| Biological Activity | Respiratory System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.