Learn More
Medchemexpress LLC Isoquinoline-d7 | 17157-12-9 | 98.0% | 136.21 | 10 MG
SDP
Catalog No. 5000454828
Encompass_Preferred
Isoquinoline-d7 is the deuterium labeled Isoquinoline. Isoquinoline is an analog of pyridine. Isoquinoline structural-based alkaloids, such as tropoloisoquinoline, phthalideisoquinoline, and naphthylisoquinoline, have anti-cancer activities. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS
Catalog No. 50-004-54828 Supplier Medchemexpress LLC Supplier No. HYW012732S10MG
Isoquinoline-d7 is the deuterium labeled Isoquinoline. Isoquinoline is an analog of pyridine. Isoquinoline structural-based alkaloids, such as tropoloisoquinoline, phthalideisoquinoline, and naphthylisoquinoline, have anti-cancer activities. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS