Learn More
Medchemexpress LLC Isoquinoline-d7 | 17157-12-9 | 98.0% | 136.21 | 100 MG
SDP
Catalog No. 5000454827
Encompass_Preferred
Isoquinoline-d7 is the deuterium labeled Isoquinoline. Isoquinoline is an analog of pyridine. Isoquinoline structural-based alkaloids, such as tropoloisoquinoline, phthalideisoquinoline, and naphthylisoquinoline, have anti-cancer activities. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-54827 Supplier Medchemexpress LLC Supplier No. HYW012732S100MG
Isoquinoline-d7 is the deuterium labeled Isoquinoline. Isoquinoline is an analog of pyridine. Isoquinoline structural-based alkaloids, such as tropoloisoquinoline, phthalideisoquinoline, and naphthylisoquinoline, have anti-cancer activities. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs