Learn More
Medchemexpress LLC Isoquinoline-d7 | 17157-12-9 | 98.0% | 136.21 | 50 MG
SDP
Catalog No. 5000454974
Encompass_Preferred
Isoquinoline-d7 is the deuterium labeled Isoquinoline. Isoquinoline is an analog of pyridine. Isoquinoline structural-based alkaloids, such as tropoloisoquinoline, phthalideisoquinoline, and naphthylisoquinoline has anti-cancer activities.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-54974 Supplier Medchemexpress LLC Supplier No. HYW012732S50MG
Isoquinoline-d7 is the deuterium labeled Isoquinoline. Isoquinoline is an analog of pyridine. Isoquinoline structural-based alkaloids, such as tropoloisoquinoline, phthalideisoquinoline, and naphthylisoquinoline has anti-cancer activities.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs