Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinoline N-Oxide, TCI America™
SDP

Catalog No. I05405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05405G Supplier TCI America Supplier No. I05405G
Add to Cart
Add to Cart

High-purity isoquinoline N-oxide building block for use as a laboratory reagent.

Chemical Identifiers

CAS 1532-72-5
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00006903
InChI Key RZIAABRFQASVSW-UHFFFAOYSA-N
Synonym isoquinoline n-oxide, isoquinoline 2-oxide, isoquinoline-n-oxide, isoquinoline oxide, isoquinoline, 2-oxide, isoquinolin-n-oxide, isoquinolin-2-ium-2-olate, isoquinolinen-oxide, acmc-1boxv, 2-oxido-isoquinolin-2-ium
PubChem CID 290378
IUPAC Name 2-oxidoisoquinolin-2-ium
SMILES C1=CC=C2C=[N+](C=CC2=C1)[O-]

Specifications

Melting Point 104°C
Quantity 5 g
Formula Weight 145.16
Percent Purity ≥98.0% (T)(qNMR)
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.