Learn More
Enzo Life Sciences Isorhamnetin (10mg). CAS: 480-19-3
SDP
Supplier:  Enzo Life Sciences ALX385024M010
Encompass_Preferred
Antiviral agent. Antioxidant. Antitumor compound. Apoptosis inducer. Alternative name: 3-Methylquercetin, 3'-Methoxy-3,4',5,7-tetrahydroxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxy-3-methoxyphenyl)-4-benzopyrone. Purity: ≥98% (HPLC). Solubility: Soluble in acetone, methanol, ethylacetate or DMSO. Only sparingly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9052