Learn More
TARGETMOL CHEMICALS INC Isorhamnetin-3-O-glucoside 5MG
SDP
Catalog No. 502880003
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Isorhamnetin-3-O-glucoside (Isorhamnetin-3-O-beta-D-Glucoside) inhibits the activity of alpha-glucosidase from rat intestine; it exhibits a potent rat lens aldose reductase (RLAR) inhibition in vitro, its IC(50) being 1.4 microM and has inhibitory effects of sorbitol accumulation, suggests that it is a leading compound for further study as a new drug for the prevention and/or treatment of diabetes and its complications. Purity 99.81%
Catalog No. 50-288-0003 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S19885MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Isorhamnetin-3-O-glucoside (Isorhamnetin-3-O-beta-D-Glucoside) inhibits the activity of alpha-glucosidase from rat intestine; it exhibits a potent rat lens aldose reductase (RLAR) inhibition in vitro, its IC(50) being 1.4 microM and has inhibitory effects of sorbitol accumulation, suggests that it is a leading compound for further study as a new drug for the prevention and/or treatment of diabetes and its complications. Purity 99.81%