Learn More
Medchemexpress LLC HY-N0776 5mg Medchemexpress, Isorhamnetin CAS:480-19-3 Purity:>98%
SDP
Catalog No. 502082822
Encompass_Preferred
Medchemexpress, HY-N0776 5mg Isorhamnetin CAS:480-19-3 Isorhamnetin is a flavonoid compound extracted from the Chinese herb Hippophae rhamnoides L.. Isorhamnetin suppresses skin cancer through direct inhibition of MEK1 and PI3K. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2822 Supplier Medchemexpress LLC Supplier No. HYN07765MG
Medchemexpress, HY-N0776 5mg Isorhamnetin CAS:480-19-3 Isorhamnetin is a flavonoid compound extracted from the Chinese herb Hippophae rhamnoides L.. Isorhamnetin suppresses skin cancer through direct inhibition of MEK1 and PI3K. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.