Learn More
Cayman Chemical IsorhamntIn-3-OD-GlucosId 10mg
SDP
Catalog No. 502910122
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; scavenges DPPH and ABTS radicals in cell-free assays (IC50s = 3.16 and 44.93 UG/ml, respectively); inhibits H. contortus egg hatching and larval development (EC50s = 431.5 and 55.61 UG/ml, respectively); induces lethality in C. elegans (EC50 = 342 UG/ml); inhibits rat lens aldose reductase (IC50 = 0.75 UG/ml) and reduces sorbitol accumulation in lenses, red blood cells, and sciatic nerve in a rat model of STZ-induced diabetes
Catalog No. 50-291-0122 Supplier Cayman Chemical Supplier No. 3560010 MG
A flavonoid glycoside with diverse biological activities; scavenges DPPH and ABTS radicals in cell-free assays (IC50s = 3.16 and 44.93 UG/ml, respectively); inhibits H. contortus egg hatching and larval development (EC50s = 431.5 and 55.61 UG/ml, respectively); induces lethality in C. elegans (EC50 = 342 UG/ml); inhibits rat lens aldose reductase (IC50 = 0.75 UG/ml) and reduces sorbitol accumulation in lenses, red blood cells, and sciatic nerve in a rat model of STZ-induced diabetes