Learn More
Medchemexpress LLC Isorhapontigenin | 32507-66-7 | MFCD12407151 | 99.8% | 258.27 | C15H14O4 | 10 MG
SDP
Catalog No. 5000288935
Encompass_Preferred
Isorhapontigenin is a dietary polyphenol (stilbenoid) research reagent that induces autophagy and exhibits anti-inflammatory and anti-tumor activity. It is intended for in vitro and preclinical research use.
  • Induces autophagy and modulates autophagy-related pathways.
  • Exhibits anti-inflammatory and anti-tumor activity in preclinical studies.
  • High purity (99.82%) suitable for research applications.
  • Offered in small solid quantities and solution formulations for experimental flexibility.
  • Chemical formula C15H14O4, molecular weight 258.27 g/mol.
Catalog No. 50-002-88935 Supplier Medchemexpress LLC Supplier No. HYN259310MG
Isorhapontigenin is a dietary polyphenol (stilbenoid) research reagent that induces autophagy and exhibits anti-inflammatory and anti-tumor activity. It is intended for in vitro and preclinical research use.
  • Induces autophagy and modulates autophagy-related pathways.
  • Exhibits anti-inflammatory and anti-tumor activity in preclinical studies.
  • High purity (99.82%) suitable for research applications.
  • Offered in small solid quantities and solution formulations for experimental flexibility.
  • Chemical formula C15H14O4, molecular weight 258.27 g/mol.