Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isorhapontigenin 96.0+%, TCI America™
SDP

Catalog No. I0804100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0804100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0804100MG Supplier TCI America Supplier No. I0804100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32507-66-7
Molecular Formula C15H14O4
Molecular Weight (g/mol) 258.273
MDL Number MFCD12407151
InChI Key ANNNBEZJTNCXHY-NSCUHMNNSA-N
Synonym 3,4′C,5-Trihydroxy-3′C-methoxy-trans-stilbene
PubChem CID 5318650
IUPAC Name 5-[(E)-2-(4-hydroxy-3-methoxyphenyl)ethenyl]benzene-1,3-diol
SMILES COC1=C(C=CC(=C1)C=CC2=CC(=CC(=C2)O)O)O

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 100 mg
Formula Weight 258.27
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isorhapontigenin
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.