Learn More
Cayman Chemical IsorhoIfolIn 10mg
SDP
Catalog No. 502950702
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; inhibits MMP-1, MMP-3, and MMP-13 (IC50s = 0.33, 2.45, and 0.22 µM, respectively); has antimutagenic properties in the E. coli WP2 tryptophan reverse mutation assay at 2 µM; active against S. aureus and P. aeruginosa in agar diffusion assays at 140 UG/ml
Catalog No. 50-295-0702 Supplier Cayman Chemical Supplier No. 3403610 MG
A flavonoid glycoside with diverse biological activities; inhibits MMP-1, MMP-3, and MMP-13 (IC50s = 0.33, 2.45, and 0.22 µM, respectively); has antimutagenic properties in the E. coli WP2 tryptophan reverse mutation assay at 2 µM; active against S. aureus and P. aeruginosa in agar diffusion assays at 140 UG/ml