Learn More
Cayman Chemical Isorhynchophylline, Purity: ≥95%, CAS: 6859-01-4, 50 mg
SDP
Catalog No. 502990072
Encompass_Preferred
An oxindole alkaloid with diverse biological activities; inhibits decreases in cell viability and increases in intracellular levels of ROS and MDA induced by Aβ (25-35) in PC12 cells; attenuates decreases in population spike amplitudes in the CA1 region of a rat hippocampal slice model of ischemia induced by oxygen and D-glucose deprivation at 30 and 100 μM; decreases the viability of HepG2, A549, BxPC-3, Caki-1, RPMI-8226, and 786-O cells (IC50s = 130-292 μM); induces apoptosis, inhibits cell migration in a wound healing assay, and decreases invasion in a Matrigel™ assay in HepG2 cells; decreases right ventricular end systolic pressure, hypertrophy, and fibrosis in a rat model of monocrotaline-induced pulmonary arterial hypertension at 0.1% in the diet
Catalog No. 50-299-0072 Supplier Cayman Chemical Supplier No. 2965050 MG
An oxindole alkaloid with diverse biological activities; inhibits decreases in cell viability and increases in intracellular levels of ROS and MDA induced by Aβ (25-35) in PC12 cells; attenuates decreases in population spike amplitudes in the CA1 region of a rat hippocampal slice model of ischemia induced by oxygen and D-glucose deprivation at 30 and 100 μM; decreases the viability of HepG2, A549, BxPC-3, Caki-1, RPMI-8226, and 786-O cells (IC50s = 130-292 μM); induces apoptosis, inhibits cell migration in a wound healing assay, and decreases invasion in a Matrigel™ assay in HepG2 cells; decreases right ventricular end systolic pressure, hypertrophy, and fibrosis in a rat model of monocrotaline-induced pulmonary arterial hypertension at 0.1% in the diet