Learn More
Medchemexpress LLC HY-N2131 5mg , Isosakuranetin CAS:480-43-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21315MG
Encompass_Preferred
HY-N2131 5mg Isosakuranetin CAS: 480-43-3 Isosakuranetin is a flavanone flavonoid which can be found in the fruit of Citrus bergamia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1949