Learn More
Cayman Chemical Isoscopoletin, Purity: ≥98%, CAS: 776-86-3, 10 mg
SDP
Catalog No. 502947256
Encompass_Preferred
A coumarin with diverse biological activities and an active metabolite of scoparone; cytotoxic to A549, MCF-7, and HepG2 cells (IC50s = 5.25, 8.58, and 4.76 µM, respectively); active against S. aureus and E. coli in agar diffusion assays; inhibits 15-LO (IC50 = 15.1 µM); reduces paw thickness in a rat model of carrageenan-induced paw edema at 0.1 mg/kg
Catalog No. 50-294-7256 Supplier Cayman Chemical Supplier No. 3411710 MG
A coumarin with diverse biological activities and an active metabolite of scoparone; cytotoxic to A549, MCF-7, and HepG2 cells (IC50s = 5.25, 8.58, and 4.76 µM, respectively); active against S. aureus and E. coli in agar diffusion assays; inhibits 15-LO (IC50 = 15.1 µM); reduces paw thickness in a rat model of carrageenan-induced paw edema at 0.1 mg/kg