Learn More
Medchemexpress LLC Isoscopoletin | 776-86-3 | 99.9% | 192.17 | C10H8O4 | 10 MG
SDP
Catalog No. 5000296383
Encompass_Preferred
Isoscopoletin is a coumarin-class small molecule provided as an analytical research standard with reported antiviral (HBV) and anti-inflammatory activity. It is supplied as a solid for in vitro and analytical applications, with high stated purity and recommended cold, light-protected storage.
  • High purity: 99.9%.
  • Solid appearance: off-white to light brown.
  • Intended for in vitro and analytical use.
  • Suitable for studies of HBV replication and signaling pathways.
  • Storage recommendation: store solid at 4°C, protect from light.
Catalog No. 50-002-96383 Supplier Medchemexpress LLC Supplier No. HYN136510MG
Isoscopoletin is a coumarin-class small molecule provided as an analytical research standard with reported antiviral (HBV) and anti-inflammatory activity. It is supplied as a solid for in vitro and analytical applications, with high stated purity and recommended cold, light-protected storage.
  • High purity: 99.9%.
  • Solid appearance: off-white to light brown.
  • Intended for in vitro and analytical use.
  • Suitable for studies of HBV replication and signaling pathways.
  • Storage recommendation: store solid at 4°C, protect from light.