Learn More
Medchemexpress LLC HY-N7043 1mg Medchemexpress, Isosilybin A CAS:142796-21-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN70431MG
Encompass_Preferred
Medchemexpress, HY-N7043 1mg Isosilybin A CAS:142796-21-2 Isosilybin A, a flavonolignan isolated from silymarin, has anti-prostate cancer (PCA) activity. Isosilybin A inhibits proliferation and induces G1 phase arrest and apoptosis in cancer cells, which activates apoptotic machinery in PCA cells via targeting Akt-NF-κB-androgen receptor (AR) axis [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3978