Learn More
Medchemexpress LLC Isosilybin B | 142796-22-3 | 5 MG
SDP
Catalog No. 5000418231
Encompass_Preferred
Isosilybin B is a flavonolignan isolated from Silybum marianum. It is known to regulate cell cycle-related proteins by reducing cyclins (D3, D1, A, E), Cdk4, Cdk2, and Cdc25A, and activating Caspases (Caspase-9 and Caspase-3). This compound promotes apoptosis, reduces androgen receptor (AR) and prostate-specific antigen (PSA), and has demonstrated anticancer activity against prostate cancer.
  • Regulates cell cycle-related proteins.
  • Activates caspases (caspase-9 and caspase-3).
  • Promotes apoptosis.
  • Reduces androgen receptor (AR) and PSA levels.
  • Exhibits anticancer activity against prostate cancer.
  • Isolated from Silybum marianum.
Catalog No. 50-004-18231 Supplier Medchemexpress LLC Supplier No. HYN70455MG
Isosilybin B is a flavonolignan isolated from Silybum marianum. It is known to regulate cell cycle-related proteins by reducing cyclins (D3, D1, A, E), Cdk4, Cdk2, and Cdc25A, and activating Caspases (Caspase-9 and Caspase-3). This compound promotes apoptosis, reduces androgen receptor (AR) and prostate-specific antigen (PSA), and has demonstrated anticancer activity against prostate cancer.
  • Regulates cell cycle-related proteins.
  • Activates caspases (caspase-9 and caspase-3).
  • Promotes apoptosis.
  • Reduces androgen receptor (AR) and PSA levels.
  • Exhibits anticancer activity against prostate cancer.
  • Isolated from Silybum marianum.