Learn More
Medchemexpress LLC Isosilybin (isosilybinin) | 72581-71-6 | 482.44 | 100 MG
SDP
Catalog No. 5000417018
Encompass_Preferred
Isosilybin (Isosilybinin) is a flavonoid derived from Silybum marianum that inhibits CYP3A4 induction with an IC50 of 74 μM. It is intended for research use only.
  • Flavonoid from Silybum marianum
  • Inhibits CYP3A4 induction with an IC50 of 74 μM
  • Stronger inhibitor of PXR-mediated CYP3A4 induction compared to silybin
  • Isosilybin B and isosilybin A exhibit anti-prostate cancer (PCA) activity through cell cycle arrest and apoptosis induction
  • Isosilybin A can activate PPARγ
Catalog No. 50-004-17018 Supplier Medchemexpress LLC Supplier No. HYN0779100MG
Isosilybin (Isosilybinin) is a flavonoid derived from Silybum marianum that inhibits CYP3A4 induction with an IC50 of 74 μM. It is intended for research use only.
  • Flavonoid from Silybum marianum
  • Inhibits CYP3A4 induction with an IC50 of 74 μM
  • Stronger inhibitor of PXR-mediated CYP3A4 induction compared to silybin
  • Isosilybin B and isosilybin A exhibit anti-prostate cancer (PCA) activity through cell cycle arrest and apoptosis induction
  • Isosilybin A can activate PPARγ