Learn More
Medchemexpress LLC Isosinensetin | 17290-70-9 | 372.37 | 20 MG
SDP
Catalog No. 5000415604
Encompass_Preferred
Isosinensetin is a flavonoid compound and an inhibitor of HIV-1 protease and PTP1B (IC50: 2.61 μM; Ki: 0.92 μM). It inhibits P-glycoprotein (P-gp) in MDR1-MDCKII cells. Isosinensetin possesses anti-tumor, anti-viral, anti-inflammatory, and antioxidant effects. It is used in research for various diseases including cancer, inflammation, osteoporosis, and diabetes.
  • Inhibits HIV-1 protease and PTP1B
  • Inhibits P-glycoprotein
  • Possesses anti-tumor, anti-viral, anti-inflammatory, and antioxidant effects
  • Used in research for cancer, inflammation, osteoporosis, and diabetes
  • Inhibits osteoclast formation and bone resorption
  • Alleviates estrogen deficiency-induced osteoporosis
  • Increases intracellular calcium ion concentration
  • Stimulates GLP-1 secretion
  • Inhibits erythrocyte hemolysis
Catalog No. 50-004-15604 Supplier Medchemexpress LLC Supplier No. HYN194120MG
Isosinensetin is a flavonoid compound and an inhibitor of HIV-1 protease and PTP1B (IC50: 2.61 μM; Ki: 0.92 μM). It inhibits P-glycoprotein (P-gp) in MDR1-MDCKII cells. Isosinensetin possesses anti-tumor, anti-viral, anti-inflammatory, and antioxidant effects. It is used in research for various diseases including cancer, inflammation, osteoporosis, and diabetes.
  • Inhibits HIV-1 protease and PTP1B
  • Inhibits P-glycoprotein
  • Possesses anti-tumor, anti-viral, anti-inflammatory, and antioxidant effects
  • Used in research for cancer, inflammation, osteoporosis, and diabetes
  • Inhibits osteoclast formation and bone resorption
  • Alleviates estrogen deficiency-induced osteoporosis
  • Increases intracellular calcium ion concentration
  • Stimulates GLP-1 secretion
  • Inhibits erythrocyte hemolysis