Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isosinensetin | 17290-70-9 | MFCD21333406 | 99.9% | 372.37 | C20H20O7 | 10 MG
SDP

Catalog No. 5000295752
Encompass_Preferred

Isosinensetin is a naturally occurring flavonoid research compound used as an inhibitor of HIV-1 protease and protein tyrosine phosphatase 1B (PTP1B), with reported activity against P-glycoprotein. It is provided as a high-purity reagent for biochemical and cell-based studies.

  • High purity (99.9%) suitable for research applications.
  • Molecular formula C20H20O7; molecular weight 372.37.
  • Reported biological activities include PTP1B and HIV-1 protease inhibition and P-gp modulation.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied as a 10 mg vial for small-scale laboratory research.

Catalog No. 50-002-95752 Supplier Medchemexpress LLC Supplier No. HYN194110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isosinensetin is a naturally occurring flavonoid research compound used as an inhibitor of HIV-1 protease and protein tyrosine phosphatase 1B (PTP1B), with reported activity against P-glycoprotein. It is provided as a high-purity reagent for biochemical and cell-based studies.

  • High purity (99.9%) suitable for research applications.
  • Molecular formula C20H20O7; molecular weight 372.37.
  • Reported biological activities include PTP1B and HIV-1 protease inhibition and P-gp modulation.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied as a 10 mg vial for small-scale laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.