Learn More
Medchemexpress LLC HY-N1941 5mg Medchemexpress, Isosinensetin CAS:17290-70-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN19415MG
Encompass_Preferred
Medchemexpress, HY-N1941 5mg Isosinensetin CAS:17290-70-9 Isosinensetin, a polymethoxylated flavone extracted from pericarpium citri reticulatae viride, exhibits inhibition on P-glycoprotein (P-gp) in MDR1-MDCKII cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2853