Learn More
Medchemexpress LLC Isosorbide | 652-67-5 | 98.8% | 25 G
SDP
Catalog No. 5000414084
Encompass_Preferred
Isosorbide is an orally active vasodilating agent. It is researched for heart failure and angina (chest pain) and also acts as an oral hyperosmotic diuretic. It is suitable for laboratory research applications.
  • Acts as an orally active vasodilating agent
  • Can be used for the research of heart failure and angina
  • Functions as an oral hyperosmotic diuretic
  • Inhibits ADP-induced platelet aggregation
  • Inhibits arachidonic acid-induced platelet aggregation
  • Increases urine volume
  • Reduces urinary concentrations of gallium and calcium
  • Contributes to fewer renal precipitates and histopathologic changes
Catalog No. 50-004-14084 Supplier Medchemexpress LLC Supplier No. 25GHYB1469
Isosorbide is an orally active vasodilating agent. It is researched for heart failure and angina (chest pain) and also acts as an oral hyperosmotic diuretic. It is suitable for laboratory research applications.
  • Acts as an orally active vasodilating agent
  • Can be used for the research of heart failure and angina
  • Functions as an oral hyperosmotic diuretic
  • Inhibits ADP-induced platelet aggregation
  • Inhibits arachidonic acid-induced platelet aggregation
  • Increases urine volume
  • Reduces urinary concentrations of gallium and calcium
  • Contributes to fewer renal precipitates and histopathologic changes