Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isosorbide | 652-67-5 | 98.8% | 500 G
SDP

Catalog No. 5000411728
Encompass_Preferred

Isosorbide is an orally active vasodilating agent used in the research of heart failure and angina (chest pain). It also acts as an oral hyperosmotic diuretic.

  • Orally active vasodilating agent
  • Used for research of heart failure and angina (chest pain)
  • Oral hyperosmotic diuretic
  • Appearance: solid, white to off-white
  • Molecular weight: 146.14
  • Formula: C6H10O4
  • Soluble in H2O and DMSO

Catalog No. 50-004-11728 Supplier Medchemexpress LLC Supplier No. HYB1469500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isosorbide is an orally active vasodilating agent used in the research of heart failure and angina (chest pain). It also acts as an oral hyperosmotic diuretic.

  • Orally active vasodilating agent
  • Used for research of heart failure and angina (chest pain)
  • Oral hyperosmotic diuretic
  • Appearance: solid, white to off-white
  • Molecular weight: 146.14
  • Formula: C6H10O4
  • Soluble in H2O and DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.