Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isosorbide Dimethyl Ether

Catalog No. 247898100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-789-8100G 100 g
24-789-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-789-8100G Supplier Sigma Aldrich Supplier No. 247898100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isosorbide dimethyl ether (DMI), a sustainable solvent, is widely used in various cosmetic and pharmaceutical formulations.General applications:DMI can be used as an alternative green solvent for epoxidation of cyclooctene, Baylis-Hillman reaction of isomaltulose-derived aldehydes with +a,+a-unsaturated ketones and also in solid-phase synthesis.It can be used as a coalescent for water-borne paints, where it acts as a co-solvent during the water loss phase and as a coalescing agent during the film formation.

Specifications

CAS 5306-85-4
Density 1.15 g/mL at 25°C
Boiling Point 93°C to 95°C (0.1 mmHg, lit.)
Molecular Formula C8H14O4
Refractive Index n20/D 1.461
Linear Formula C8H14O4
MDL Number MFCD00064828
Quantity 100 g
Synonym DMI; Dimethyl isosorbide; 1,4:3,6-Dianhydro-2,5-di-O-methyl-D-glucitol; 1,4:3,6-Dianhydrosorbitol 2,5-dimethyl ether; 2,5-Di-O-methyl-1,4:3,6-dianhydro-D-glucitol
Molecular Weight (g/mol) 174.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.